Learn More
Heidolph™ Viton™ Tubing
Shop All Heidolph Instruments Products
Click to view available options
Tubing Size:
0.8
1.7
7.9
Thickness (Metric):
1.6 mm
2.5 mm
Unit Size:
1m
Each
Description
- Low gas permeability.
- Resistant to solvents and corrosives at high temperatures.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Borosilicate Glass |
| Thickness (Metric) | 1.6 mm |
| Sterility | Non-sterile |
| Tubing Size | 0.8 |
| Maximum Pressure | 0.7/1/7 bar |