Learn More
MP Biomedicals Vitamin B Complex Diet, Complete, Modified, Powder
Product Code. 11482871 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 kg
20 kg
50 kg
Unit Size:
10kg
20kg
50kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482871 10 kg 10kg
11492871 20 kg 20kg
11402881 50 kg 50kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11482871 Supplier MP Biomedicals Supplier No. 0296017010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Our traditional vitamin B complex diet, complete, but revised to include essential trace minerals, DL-methionine and fiber.
Our traditional vitamin B complex diet, complete, but revised to include essential trace minerals, DL-methionine and fiber.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Product Type Animal Diet
Disposable Single-use
Format Powder
Quantity 10 kg
Sterility Non-sterile
Storage Requirements Room Temperature
Length (English) 1 ft.
Length (Metric) 0.3048 m
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.