Learn More
VistaLab™ Technologies Reagent Reservoirs: 12-Channel
Product Code. 16485094
Click to view available options
Sterility:
Non-sterile
Sterile
Packaging:
Bulk
Individually Wrapped
Quantity:
10/Cs.
25/Cs.
Unit Size:
Pack of 10
Pack of 25
Product Code. 16485094
Brand: VistaLab™ Technologies 40541010
Unique trough-within-trough design maximizes reagent recovery.
  • Polystyrene with trimmed base corners
  • Distinct pour-off spouts to empty reservoirs without spills
  • Well-marked graduated increments on inside wall minimize reagent waste
  • Wide base and sturdy construction provide stability and help to avoid spills
  • Molded corner structures allow easy separation of bulk packaged reservoirs
  • 100mL model (Cat. No. 21-382-111) supplied with lid
  • Available individually wrapped or bulk packaged, gamma sterilized or nonsterile
  • 5 mL capacity / well
Specifications
Certifications/Compliance RNase/DNase, Pyrogen-free
Color Clear
Capacity (Metric) 60 mL
Material Polypropylene
Sterility Non-sterile
Type Reagent Reservoir
For Use With (Application) General Liquid Handling
Height (English) 5 in.
Includes Lid
Length (English) 12 in.
Packaging Bulk
Quantity 10/Cs.
Volume (Metric) Well 5 mL
Width (English) 6 in.
Autoclavable Autoclavable
Channels 12
Disposable Single-use
Bottom Shape V-bottom