missing translation for 'onlineSavingsMsg'
Learn More

Visomitin, MedChemExpress

Product Code. 16429606
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16429606 50 mg 50mg
16472586 25 mg 25mg
16453038 10 mg 10mg
16425777 5 mg 5mg
16404346 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16429606 Supplier Medchem Express Supplier No. HY100474

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Visomitin (SKQ1) is a mitochondrial-targeted antioxidant with the high mitochondrion membrane penetrating ability and potent antioxidant capability.

Visomitin (SKQ1) is a mitochondrial-targeted antioxidant with the high mitochondrion membrane penetrating ability and potent antioxidant capability.

Specifications

Chemical Name or Material Visomitin
Physical Form Solid
CAS 934826-68-3
Color Brownish Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C36H42BrO2P
For Use With (Application) COVID-19-immunoregulation
Synonym SKQ1
Solubility Information DMSO : 100 mg/mL (161.92 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (80.96 mM; Need ultrasonic) ∣H2O : 3.33 mg/mL (5.39 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(C(CCCCCCCCCC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3)=C4)C(C)=C(C)C4=O.[Br-]
Molecular Weight (g/mol) 617.6
Formula Weight 617.6
Percent Purity 95.57%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.