missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Visocolor™ HE Colorimetric Test Kit Refill Pack

Product Code. 10641363 Shop All Macherey-Nagel Products
Change view
Click to view available options
No. of Tests:
120
Unit Size:
1 test
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests unitSize
10641363 120 1 test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10641363 Supplier Macherey-Nagel™ Supplier No. 920187

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Replace consumed chemicals from test kits. Macherey-Nagel™ Visocolor™ HE Colorimetric Test Kit Refill Packs are reagents used with HE tests to determine even the lowest limiting values. The kits use longer measuring tubes and larger sample volumes for the highest sensitivity and accuracy.

Visocolor™ HE test kits provide highly precise analysis via exact dosing of the single reagents as well as compensating for turbidity and color. The sensitivity of Visocolor™ HE is 10 to 100 times higher compared to other Visocolor tests.

  • Visual evaluation
  • Environment-friendly, without toxic reagents
  • Economically priced
  • Convenient handling
  • Highest accuracy due to extremely narrow gradation
  • Highest sensitivity down to 0.002mg/L due to longer measuring tubes
  • Reagent bottles with clear dosing instructions
  • Compensation of turbidities and colors
  • Refill packs available

Specifications

Analytes Silica (Si4+)
Color Color change: White to Blue
Description Highly sensitive
Format Liquid Reagent
For Use With (Application) Silicon determination
For Use With (Equipment) HE Colorimetric Test Kit, Silica (mfr. no. 920 087)
Graduations 0.0 - 0.01 - 0.02 - 0.03 - 0.05 - 0.07 - 0.10 - 0.15 - 0.20 - 0.30mg/L Si
Includes Reagent bottles
No. of Tests 120
Quantity 1 Each
Range 0 to 0.30mg/L
Test Method Silico molybdenum blue
Test Time 9 min.
Type Reagent set
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • VISOCOLOR HE Silicon, refill pack
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: sulfuric acid, ammonium heptamolybdate,sodium disulfite, oxalic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.