Learn More
(+)-Viridiflorol, TRC
Product Code. 30138302
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138302 100 mg 100mg
30008747 25 mg 25mg
30009400 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138302 Supplier Toronto Research Chemicals Supplier No. TRCV673900100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 552-02-3
Molecular Formula C15H26O
Molecular Weight (g/mol) 222.37
Synonym (1aR,4S,4aS,7R,7aS,7bS)-Decahydro-1,1,4,7-tetramethyl-1H-cycloprop[e]azulen-4-ol, Himbaccol, [1aR-(1aα,4β,4aβ,7α,7aβ,7bα)]Decahydro-1,1,4,7-tetramethyl-1H-cycloprop[e]azulen-4-ol, d-Viridiflorol;
IUPAC Name (1aR,4S,4aS,7R,7aS,7bS)-1,1,4,7-tetramethyl-2,3,4a,5,6,7,7a,7b-octahydro-1aH-cyclopropa[e]azulen-4-ol
SMILES C[C@@H]1CC[C@H]2[C@@H]1[C@H]3[C@H](C3(C)C)CC[C@]2(C)O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C15H26O/c1-9-5-6-10-12(9)13-11(14(13,2)3)7-8-15(10,4)16/h9-13,16H,5-8H2,1-4H3/t9-,10+,11-,12-,13-,15+/m1/s1
Formula Weight 222.1984
Percent Purity >95%
Chemical Name or Material (+)-Viridiflorol
Grade GC