missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VIP36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590437
This item is not returnable.
View return policy
Description
Positive Samples: Mouse brain, Mouse heart, Mouse kidney; Cellular Location: Endoplasmic reticulum membrane, Endoplasmic reticulum-Golgi intermediate compartment membrane, Golgi apparatus membrane, Single-pass membrane protein, Single-pass type I membrane protein.
This gene encodes a type I transmembrane lectin that shuttles between the endoplasmic reticulum, the Golgi apparatus and the plasma membrane. The encoded protein binds high mannose type glycoproteins and may facilitate their sorting, trafficking and quality control.
Specifications
| VIP36 | |
| Polyclonal | |
| Unconjugated | |
| LMAN2 | |
| 1110003H06Rik; 1300009F09Rik; AA408240; AL023023; AU040819; C5orf8; glycoprotein GP36b; GP36B; Lectin mannose-binding 2; lectin, mannose binding 2; lectin, mannose-binding 2; Lman2; vesicular integral protein of 36 kDa; Vesicular integral-membrane protein 36; Vesicular integral-membrane protein VIP36; Vip36; VIP36 (vesicular integral-membrane protein); wu:fb16f04; zgc:158761 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 66890 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.35 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9DBH5 | |
| LMAN2 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction