missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This gene is a member of the visinin/recoverin subfamily of neuronal calcium sensor proteins. The encoded protein is strongly expressed in granule cells of the cerebellum where it associates with membranes in a calcium-dependent manner and modulates intracellular signaling pathways of the central nervous system by directly or indirectly regulating the activity of adenylyl cyclase. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.
Specifications
Specifications
| Antigen | VILIP-1 |
| Applications | Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.63 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | VSNL1 |
| Gene Accession No. | P62760, P62761, P62762 |
| Gene Alias | Hippocalcin like protein 3, HLP3, HPCAL3, HUVISL1, VILIP, VILIP 1, VILIP1, visinin like 1, Visinin like protein 1, VISL1, VLP 1, VLP1, VLP-1, VSNL1 |
| Gene Symbols | Vsnl1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?