Learn More
Glycidyl Eicosapentaenoate-d5, TRC
Product Code. 30240794
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30240794 10 mg 10mg
30240835 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30240794 Supplier Toronto Research Chemicals Supplier No. TRCG61565210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1795143-63-3
Formula Weight 363.2822
InChI Formula InChI=1S/C23H34O3/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-23(24)26-21-22-20-25-22/h3-4,6-7,9-10,12-13,15-16,22H,2,5,8,11,14,17-21H2,1H3/b4-3-,7-6-,10-9-,13-12-,16-15-/i20D2,21D2,22D
Molecular Formula C23H29D5O3
Molecular Weight (g/mol) 363.55
SMILES O=C(OC([2H])([2H])C1([2H])OC1([2H])[2H])CCC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CC
Synonym (all-Z)-5,8,11,14,17-Eicosapentaenoic Acid Glycidyl-d5 Ester, EPA Glycidyl-d5 Ester;
Chemical Name or Material Glycidyl Eicosapentaenoate-d5
Specifications
Quantity 10 mg