Learn More
Invitrogen™ Human DNAH2 (aa 3776-3852) Control Fragment Recombinant Protein
Product Code. 30201631
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 30201631
Brand: Invitrogen™ RP103886
Recombinant Protein
Highest antigen sequence indentity to the following orthologs: Mouse (96%), Rat (96%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64309 (PA5-64309. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Force generating protein of respiratory cilia. Produces force towards the minus ends of microtubules. Dynein has ATPase activity; the force-producing power stroke is thought to occur on release of ADP. Involved in sperm motility; implicated in sperm flagellar assembly.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number Q9P225
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 146754
Name Human DNAH2 (aa 3776-3852) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias axonemal beta dynein heavy chain 2; Ciliary dynein heavy chain 2; DNAH2; DNAHC2; DNHD3; dynein axonemal heavy chain 2; dynein heavy chain 2, axonemal; Dynein heavy chain domain-containing protein 3; dynein, axonemal, heavy chain 2; dynein, axonemal, heavy polypeptide 2; KIAA1503
Common Name DNAH2
Gene Symbol DNAH2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PGEWENACNEMQRMLIVRSLRQDRVAFCVTSFIITNLGSRFIEPPVLNMKSVLEDSTPRSPLVFILSPGVDPTSALL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining