Learn More
R&D Systems™ Recombinant Mouse TfR (Transferrin R) Protein
Product Code. 18756139
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Product Code. 18756139
Brand: R&D Systems™ 9706TR050
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Measured by its binding ability in a functional ELISA. When Recombinant Mouse TfR (Transferrin R) (Catalog # 9706-TR) is immobilized at 5 μg/mL (100 μL/well), Biotinylated Mouse Holo-Transferrin binds with an ED50 of 20.0-160 ng/mL.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number Q62351
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 7037
Molecular Weight (g/mol) Observed M.W.: 75-86 kDa (major) and 133-157 kDa (minor), under reducing conditions.
Name CD71, CD71 antigen, CD71TRFR, p90, T9, TfR, TfR (TransferrinR), TFR1, TFRC, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse TfR (Transferrin R) protein Cys89-Phe763,with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 500μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.