Learn More
Human ChemR23 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB362-SP
162.35 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
ChemR23/CMKLR1 Monoclonal specifically detects ChemR23/CMKLR1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| ChemR23/CMKLR1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CHEMERINR, chemokine receptor-like 1, chemokine-like receptor 1, CHEMR23, cmklr1, Dez, Gpcr27, G-protein coupled receptor ChemR23, G-protein coupled receptor DEZ, MGC126105, MGC126106, orphan G-protein coupled receptor, Dez | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1240 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG3 |
| Flow Cytometry, CyTOF | |
| 84939 | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| NP_004063 | |
| CMKLR1 | |
| NS0 mouse myeloma cell line transfected with human ChemR23 Met1-Leu371 Accession # NP_004063 | |
| 25 μg | |
| Primary | |
| Detects human ChemR23. Stains human ChemR23-transfected cells but not irrelevant transfectants. | |
| Human | |
| Purified |
Product Content Correction
Product Title
For Research Use Only