Learn More
Promed Bioscience Atelocollagen, Porcine Type I (Enzymatic extraction)
Click to view available options
Quantity:
10 mg
20 mg
Unit Size:
10mg
20mg
Description
- Lyophilized, highly purified Type I atelocollagen with superior functionality and physical/mechanical properties (such as tensile strength and elasticity)
- Proprietary enzyme-based extraction removes telopeptides at the non-helical N and C termini, resulting in a triple helix configuration that exhibits reduced immunogenicity and enhanced biocompatibility
- Suitable for in vitro cell cultures, for cell functional assays, as a chemoattractant agent and for soft 3D cell cultures (e.g. tumor assays)
Specifications
Specifications
| Formulation | Lyophilized Fibrous Powder |
| Purification Method | Dialysis |
| Quantity | 10 mg |
| Content And Storage | -20°C to 8°C |
| Endotoxin Level | <1.0 EU/mL |
| Purity or Quality Grade | >95% (by SDS/Western blotting) |