Learn More
SCAT Safety Waste Set V2.0
Product Code. 16679085 Shop All SCAT Europe Products
Click to view available options
Volume (Metric):
10 L Canister
2.5 L Canister
5 L Canister
Includes:
Safety Funnel with Lid 'Arnold'
Safety funnel with lid 'MARCO'
Unit Size:
1 set
Each
Product Code. 16679085
Brand: SCAT 307318
Only null left
Add to basket
Add to basket
Safety Waste Set V2.0 includes everything need to get your waste fluid collection site ready for operation quickly.
  • Collect liquid waste safely with the SCAT. safety funnel MARCO V2.0
  • Ready to use
  • Consists of lid funnel and different canister sizes in a complete set
Specifications
Diameter (Metric) Inner 140 mm
Length (Metric) 200 mm
Includes Safety funnel with lid 'MARCO'
Material HDPE
Thread Size GL45
Volume (Metric) 10 L Canister
Type Waste Funnel