Learn More
Invitrogen™ CEACAM21 Polyclonal Antibody
GREENER_CHOICE
Product Code. 16667974
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16667974 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16667974 Supplier Invitrogen™ Supplier No. PA5101499
Rabbit Polyclonal Antibody
Antibody detects endogenous levels of total CEACAM21.
CEACAM21 (Carcinoembryonic Antigen Related Cell Adhesion Molecule 21) is a Protein Coding gene. An important paralog of this gene is CEACAM1.
TRUSTED_SUSTAINABILITY
Specifications
Antigen CEACAM21
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CEACAM21
Gene Accession No. Q3KPI0
Gene Alias carcinoembryonic antigen related cell adhesion molecule 21; carcinoembryonic antigen-related cell adhesion molecule 21; CEACAM21; CEACAM3; R29124_1; UNQ3098/PRO10075
Gene Symbols CEACAM21
Host Species Rabbit
Immunogen A synthesized peptide derived from human CEACAM21(Accession Q3KPI0), corresponding to amino acid residues L246-S293.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90273
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG