Learn More
Diquat dibromid Monohydrate, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
Specifications
Specifications
| Chemical Name or Material | Diquat dibromid Monohydrate |
| CAS | 6385-62-2 |
| CAS Max % | ≤ 100% |
| Linear Formula | C12H12Br2N2 · H2O |
| Beilstein | 3769318 |
| Color | Colorless to Yellow |
| Melting Point | 325°C |
| Synonym | 9,10-dihydro-8a,10a-diazoniaphenanthrene dibromide; 1,1'-ethylene-2,2'-bipyridyldiylium dibromide |
| Molecular Weight (g/mol) | 362.06 g/mol |
| Quantity | 100 mg |