Learn More
RIG-1 modulator 1, MedChemExpress
Product Code. 16491089
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
Product Code. 16491089
Brand: Medchem Express HY107902
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress RIG-1 modulator 1 is an anti-viral compound which can be useful for the treatment of viral infections including influenza virus, HBV, HCV and HIV extracted from patent WO 2015172099 A1.
RIG-1 modulator 1 is an anti-viral compound which can be useful for the treatment of viral infections including influenza virus, HBV, HCV and HIV extracted from patent WO 2015172099 A1.
Specifications
Chemical Name or Material RIG-1 modulator 1
Physical Form Solid
CAS 1428729-63-8
Color Light Yellow
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C14H17N5OS2
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : ≥ 35 mg/mL (104.34 mM)
SMILES O=C(NC1=NC2=CC=C(SC(C)=N3)C3=C2S1)NCCN(C)C
Molecular Weight (g/mol) 335.45
Formula Weight 335.45
Percent Purity 98.01%
Grade Research