Learn More
(+)-JQ-1, MedChemExpress
Product Code. 16488207
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5mg
Product Code. 16488207
Brand: Medchem Express HY13030
MedChemExpress (+)-JQ-1 (JQ1) is a potent, specific, and reversible BET bromodomain inhibitor, with IC50s of 77 and 33 nM for the first and second bromodomain (BRD4(1/2)). (+)-JQ-1 also activates autophagy.
(+)-JQ-1 (JQ1) is a potent, specific, and reversible BET bromodomain inhibitor, with IC50s of 77 and 33 nM for the first and second bromodomain (BRD4(1/2)). (+)-JQ-1 also activates autophagy.
Specifications
Chemical Name or Material (+)-JQ-1
CAS 1268524-70-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H25ClN4O2S
Quantity 5 mg
Synonym JQ1
Solubility Information DMSO : ≥ 45 mg/mL (98.47 mM)
SMILES O=C(C[C@H]1C2=NN=C(N2C3=C(C(C4=CC=C(C=C4)Cl)=N1)C(C)=C(S3)C)C)OC(C)(C)C
Molecular Weight (g/mol) 456.99
Formula Weight 456.99
Percent Purity 98.65%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death