Learn More
Hesperin, MedChemExpress
Product Code. 16483338
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16483338
Brand: Medchem Express HY101371
MedChemExpress Hesperin is a bioactive ingredient present in Japanese horseradish (wasabi) and has been shown to be an Nrf2 activator.
Hesperin is a bioactive ingredient present in Japanese horseradish (wasabi) and has been shown to be an Nrf2 activator.
Specifications
Chemical Name or Material Hesperin
Physical Form Liquid
CAS 4430-35-7
Color Colorless
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C8H15NOS2
For Use With (Application) Cancer-programmed cell death
Solubility Information Ethanol : 100 mg/mL (487.00 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (243.50 mM; Need ultrasonic)
SMILES O=S(CCCCCCN=C=S)C
Molecular Weight (g/mol) 205.34
Formula Weight 205.34
Percent Purity 98.12%
Grade Research