Learn More
Caftaric acid, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Caftaric acid is a natural product.
Specifications
Specifications
| Chemical Name or Material | Caftaric acid |
| CAS | 67879-58-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C13H12O9 |
| Quantity | 5 mg |
| Synonym | trans-Caftaric acid |
| Solubility Information | H2O : 50 mg/mL (160.14 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (160.14 mM; Need ultrasonic) |
| SMILES | O=C(O)[C@H](OC(/C=C/C1=CC=C(O)C(O)=C1)=O)[C@@H](O)C(O)=O |
| Molecular Weight (g/mol) | 312.23 |
| Show More |