Learn More
JQEZ5, MedChemExpress
Product Code. 16479736
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16479736
Brand: Medchem Express HY100846
MedChemExpress JQEZ5 is a potent and selective EZH2 lysine methyltransferase inhibitor. JQEZ5 SAM-competitive inhibition of polycomb repressive complex 2 (PRC2) with an IC50 of 80 nM. JQEZ5 has anti-tumor effects.
JQEZ5 is a potent and selective EZH2 lysine methyltransferase inhibitor. JQEZ5 SAM-competitive inhibition of polycomb repressive complex 2 (PRC2) with an IC50 of 80 nM. JQEZ5 has anti-tumor effects.
Specifications
Chemical Name or Material JQEZ5
CAS 1913252-04-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C30H38N8O2
Quantity 50 mg
Solubility Information DMSO : 25 mg/mL (46.07 mM; Need ultrasonic)
SMILES CC(N1N=CC2=C(C(NCC3=C(C=C(NC3=O)C)CCC)=O)C=C(C4=CC=C(N=C4)N5CCN(C)CC5)N=C12)C
Molecular Weight (g/mol) 542.68
Formula Weight 542.68
Percent Purity 98.19%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death