Learn More
D159687, MedChemExpress
Product Code. 16479107
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16479107
Brand: Medchem Express HY15444
MedChemExpress D159687 is a selective PDE4D inhibitor.
D159687 is a selective PDE4D inhibitor.
Specifications
Chemical Name or Material D159687
CAS 1155877-97-6
Color White
Purity Grade Notes Research
Molecular Formula C21H19ClN2O2
Quantity 5 mg
Solubility Information DMSO : 150 mg/mL (408.90 mM; Need ultrasonic)
SMILES NC(NC1=CC=C(CC2=CC(C3=CC(Cl)=CC=C3)=C(OC)C=C2)C=C1)=O
Molecular Weight (g/mol) 366.84
Formula Weight 366.84
Percent Purity 98.12%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation