Learn More
STO-609, MedChemExpress
Product Code. 16473857
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16473857
Brand: Medchem Express HY19805
MedChemExpress STO-609 is a selective and cell-permeable inhibitor of the Ca2+/calmodulin-dependent protein kinase kinase (CaM-KK), with Ki values of 80 and 15 ng/mL for recombinant CaM-KKα and CaM-KKβ, respectively. STO-609 inhibits AMP-activated protein kinase kinase (AMPKK) activity in HeLa cell lysates with an IC50 ∼0.02 g/ml.
STO-609 is a selective and cell-permeable inhibitor of the Ca2+/calmodulin-dependent protein kinase kinase (CaM-KK), with Ki values of 80 and 15 ng/mL for recombinant CaM-KKα and CaM-KKβ, respectively. STO-609 inhibits AMP-activated protein kinase kinase (AMPKK) activity in HeLa cell lysates with an IC50 ∼0.02 g/ml.
Specifications
Chemical Name or Material STO-609
CAS 52029-86-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H10N2O3
Quantity 50 mg
Solubility Information DMSO : 6.25 mg/mL (19.89 mM; ultrasonic and warming and heat to 80°C) ∣DMF : 5.56 mg/mL (17.69 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=C2C3=C(C4=NC5=CC=CC=C5N4C(C3=CC=C2)=O)C=C1)O
Molecular Weight (g/mol) 314.29
Formula Weight 314.29
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism