Learn More
Thermo Scientific™ Store Image and Database Management Software, standard edition
Click to view available options
Quantity:
1 each
Unit Size:
Each
Description
Using Thermo Scientific™ Store™ Informatics Software, store and manage data automatically for rapid, highly scalable, global access to all HCS data.
Thermo Scientific™ Store™ Image and Database Management Software is a SQL Server or Oracle™database used for storing and managing the images and data automatically transferred from an HCS instrument or imported from another imaging source. It allows for rapid, highly scalable, global access to both data and images. Use the Store Image and Database Management Software after you have collected data and images.
Highlights:
Highlights:
- Automatic Transfer of Images and Data – Centralized data storage allowing data and image access from multiple locations
- Accessible and Linked – Data is seamlessly linked to its associated image
- Visualize – View images and data linked together and easily compare images
- Export – Built in export formats for software such as Spotfire™, Excel™, and GeneData Screener™ programs
- HCS Connect – Fully open standard, web services API that provides programmatic access to any data and images being managed by Store software
- HCS Gateway – Users can then benefit from a single, open standard repository of images and data regardless of source
Specifications
Specifications
| Description | Store Informatics Software Standard Edition |
| For Use With (Equipment) | ArrayScan High Content Analysis (HCA) Readers, CellInsight CX5 High Content Screening (HCS) Platform |
| Software | Image Management, Database Management |
| License | Standard |
| Quantity | 1 each |
For Research Use Only. Not for use in diagnostic procedures.