Learn More
Asapiprant, MedChemExpress
Product Code. 16447176
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Product Code. 16447176
Brand: Medchem Express HY16763
MedChemExpress Asapiprant is a potent and selective DP1 receptor antagonist with a Ki of 0.44 nM.
Asapiprant is a potent and selective DP1 receptor antagonist with a Ki of 0.44 nM.
Specifications
Chemical Name or Material Asapiprant
CAS 932372-01-5
Color White
Purity Grade Notes Research
Molecular Formula C24H27N3O7S
Quantity 100 mg
Synonym S-555739
Solubility Information DMSO : 11.11 mg/mL (22.15 mM; Need ultrasonic)
SMILES O=C(O)COC1=CC(N2CCN(S(=O)(C3=CC=C(OC(C)C)C=C3)=O)CC2)=CC=C1C4=NC=CO4
Molecular Weight (g/mol) 501.55
Formula Weight 501.55
Percent Purity 99.4%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation