Learn More
(S)-JQ-35, MedChemExpress
Product Code. 16445656
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16445656
Brand: Medchem Express HY117286
MedChemExpress (S)-JQ-35 (TEN-010) is an inhibitor of the Bromodomain and Extra-Terminal (BET) family bromodomain-containing proteins with potential antineoplastic activity.
(S)-JQ-35 (TEN-010) is an inhibitor of the Bromodomain and Extra-Terminal (BET) family bromodomain-containing proteins with potential antineoplastic activity.
Specifications
Chemical Name or Material (S)-JQ-35
Physical Form Solid
CAS 1349719-98-7
Color Pink
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C27H34ClN7OS
For Use With (Application) Cancer-programmed cell death
Synonym TEN-010
Solubility Information DMSO : ≥ 85 mg/mL (157.37 mM)
SMILES O=C(NCCCN1CCN(C)CC1)C[C@H]2C3=NN=C(C)N3C4=C(C(C)=C(C)S4)C(C5=CC=C(Cl)C=C5)=N2
Molecular Weight (g/mol) 540.12
Formula Weight 540.12
Percent Purity 95.99%
Grade Research