Learn More
HC-070, MedChemExpress
Product Code. 16440917
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16440917
Brand: Medchem Express HY112302
MedChemExpress HC-070 is an antagonist of TRPC4/TRPC5, with IC50s of 9.3 nM and 46 nM for hTRPC5 and hTRPC4 in cells, respectively.
HC-070 is an antagonist of TRPC4/TRPC5, with IC50s of 9.3 nM and 46 nM for hTRPC5 and hTRPC4 in cells, respectively.
Specifications
Chemical Name or Material HC-070
CAS 1628291-95-1
Color White
Purity Grade Notes Research
Molecular Formula C22H20Cl2N4O4
Quantity 50 mg
Solubility Information DMSO : ≥ 62.5 mg/mL (131.49 mM)
SMILES O=C(N1CCCO)N(C)C2=C(N(CC3=CC=C(Cl)C=C3)C(OC4=CC=CC(Cl)=C4)=N2)C1=O
Molecular Weight (g/mol) 475.32
Formula Weight 475.32
Percent Purity 98.39%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation