Learn More
CA-170, MedChemExpress
Product Code. 16435987
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
Product Code. 16435987
Brand: Medchem Express HY101093
MedChemExpress CA-170 is an orally delivered dual inhibitor of VISTA and PD-L1. CA-170 exhibits potent rescue of proliferation and effector functions of T cells inhibited by PD-L1/L2 and VISTA with selectivity over other immune checkpoint proteins as well as a broad panel of receptors and enzymes.
CA-170 is an orally delivered dual inhibitor of VISTA and PD-L1. CA-170 exhibits potent rescue of proliferation and effector functions of T cells inhibited by PD-L1/L2 and VISTA with selectivity over other immune checkpoint proteins as well as a broad panel of receptors and enzymes.
Specifications
Chemical Name or Material CA-170
Physical Form Solid
CAS 1673534-76-3
Color Light Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C12H20N6O7
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 28.5 mg/mL (79.10 mM; Need ultrasonic and warming)
SMILES C[C@@H](O)[C@@H](C(O)=O)NC(N[C@H](C1=NC([C@@H](N)CO)=NO1)CC(N)=O)=O
Molecular Weight (g/mol) 360.32
Formula Weight 360.32
Percent Purity 95.14%
Grade Research