Learn More
ChX710, MedChemExpress
Product Code. 16431027
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16431027
Brand: Medchem Express HY112951
MedChemExpress ChX710 could prime the type I interferon response to cytosolic DNA, which induces the ISRE promoter sequence, specific cellular Interferon-Stimulated Genes (ISGs), and the phosphorylation of Interferon Regulatory Factor (IRF) 3.
ChX710 could prime the type I interferon response to cytosolic DNA, which induces the ISRE promoter sequence, specific cellular Interferon-Stimulated Genes (ISGs), and the phosphorylation of Interferon Regulatory Factor (IRF) 3.
Specifications
Chemical Name or Material ChX710
Physical Form Solid
CAS 2438721-44-7
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H21N5O
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 20 mg/mL (61.84 mM; Need ultrasonic)
SMILES O=C(NC(C)CN(C)C)C1=C(N=C(C2=CC=NC=C2)N3)C3=CC=C1
Molecular Weight (g/mol) 323.39
Formula Weight 323.39
Percent Purity 99.12%
Grade Research