Learn More
PH-797804, MedChemExpress
Product Code. 16420986
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5mg
Product Code. 16420986
Brand: Medchem Express HY10403
MedChemExpress PH-797804 is a ATP-competitive, selective p38α/p38β inhibitor (IC50=26 nM and Ki=5.8 nM for p38α; Ki=40 nM for p38β) and does not inhibit JNK2.
PH-797804 is a ATP-competitive, selective p38α/p38β inhibitor (IC50=26 nM and Ki=5.8 nM for p38α
Specifications
Chemical Name or Material PH-797804
CAS 586379-66-0
Color White
Purity Grade Notes Research
Molecular Formula C22H19BrF2N2O3
Quantity 200 mg
Solubility Information DMSO : 50 mg/mL (104.76 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C(N2C(C)=CC(OCC3=CC=C(C=C3F)F)=C(C2=O)Br)=C1)C)NC
Molecular Weight (g/mol) 477.3
Formula Weight 477.3
Percent Purity 98.94%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation