Learn More
Cilofexor, MedChemExpress
Product Code. 16415206
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16415206
Brand: Medchem Express HY109083
MedChemExpress Cilofexor (GS-9674) is a potent, selective and orally active nonsteroidal FXR agonist with an EC50 of 43 nM. Cilofexor has anti-inflammatory and antifibrotic effects. Cilofexor has the potential for primary sclerosing cholangitis (PSC) and nonalcoholic steatohepatitis (NASH) research.
Cilofexor (GS-9674) is a potent, selective and orally active nonsteroidal FXR agonist with an EC50 of 43 nM. Cilofexor has anti-inflammatory and antifibrotic effects. Cilofexor has the potential for primary sclerosing cholangitis (PSC) and nonalcoholic steatohepatitis (NASH) research.
Specifications
Chemical Name or Material Cilofexor
CAS 1418274-28-8
Color White
Purity Grade Notes Research
Molecular Formula C28H22Cl3N3O5
Quantity 100 mg
Synonym GS-9674
Solubility Information DMSO : 50 mg/mL (85.20 mM; Need ultrasonic)
SMILES ClC1=C(C2(CN(C3=CC(C(O)=O)=CC=N3)C2)O)C=CC(OCC4=C(C5CC5)ON=C4C(C(Cl)=CC=C6)=C6Cl)=C1
Molecular Weight (g/mol) 586.85
Formula Weight 586.85
Percent Purity 98.25%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation