Learn More
NGP555, MedChemExpress
Product Code. 16414238
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16414238
Brand: Medchem Express HY108714
MedChemExpress NGP555 is a γ-secretase modulator.
NGP555 is a γ-secretase modulator.
Specifications
Chemical Name or Material NGP555
CAS 1304630-27-0
Color White
Purity Grade Notes Research
Molecular Formula C23H23FN4S
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (61.50 mM; Need ultrasonic)
SMILES CC1=CN(C2=CC=C(C3=CSC(NC4=CC(CC)=C(C)C=C4C)=N3)C=C2F)C=N1
Molecular Weight (g/mol) 406.52
Formula Weight 406.52
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration