Learn More
3-Deazaneplanocin A hydrochloride, MedChemExpress
Product Code. 16413286
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
Product Code. 16413286
Brand: Medchem Express HY12186
MedChemExpress 3-Deazaneplanocin A hydrochloride (DZNep hydrochloride) is a potent histone methyltransferase EZH2 inhibitor. 3-Deazaneplanocin A hydrochlorideis a potent S-adenosylhomocysteine hydrolase (AHCY) inhibitor. 3-Deazaneplanocin A hydrochloride also has anti-orthopoxvirus and anti-variola activities.
3-Deazaneplanocin A hydrochloride (DZNep hydrochloride) is a potent histone methyltransferase EZH2 inhibitor. 3-Deazaneplanocin A hydrochlorideis a potent S-adenosylhomocysteine hydrolase (AHCY) inhibitor. 3-Deazaneplanocin A hydrochloride also has anti-orthopoxvirus and anti-variola activities.
Specifications
Chemical Name or Material 3-Deazaneplanocin A hydrochloride
CAS 120964-45-6
Color Light Brown
Purity Grade Notes Research
Molecular Formula C12H15ClN4O3
Quantity 25 mg
Synonym DZNep hydrochloride NSC 617989 hydrochloride 3-Deazaneplanocin hydrochloride
Solubility Information H2O : 50 mg/mL (167.38 mM; Need ultrasonic)
SMILES O[C@@H]1[C@H](O)C(CO)=C[C@H]1N2C=NC3=C2C=CN=C3N.[H]Cl
Molecular Weight (g/mol) 298.73
Formula Weight 298.73
Percent Purity 98.37%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death