Learn More
LJI308, MedChemExpress
Product Code. 16400328
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16400328
Brand: Medchem Express HY19713
MedChemExpress LJI308 is a potent pan-ribosomal S6 kinase (RSK) inhibitor, with IC50s of 6 nM, 4 nM, and 13 nM for RSK1, RSK2, and RSK3, respectively. LJI308 inhibits the phosphorylation of RSK (T359/S363) and YB-1 (S102) after irradiation, treatment with EGF, and in cells expressing a KRAS mutation.
LJI308 is a potent pan-ribosomal S6 kinase (RSK) inhibitor, with IC50s of 6 nM, 4 nM, and 13 nM for RSK1, RSK2, and RSK3, respectively. LJI308 inhibits the phosphorylation of RSK (T359/S363) and YB-1 (S102) after irradiation, treatment with EGF, and in cells expressing a KRAS mutation.
Specifications
Chemical Name or Material LJI308
CAS 1627709-94-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H18F2N2O2
Quantity 5 mg
Solubility Information DMSO : 25 mg/mL (67.86 mM; Need ultrasonic)
SMILES OC1=C(F)C=C(C2=C(C3=CC=C(N4CCOCC4)C=C3)C=CN=C2)C=C1F
Molecular Weight (g/mol) 368.38
Formula Weight 368.38
Percent Purity 99.21%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease