Learn More
Cole-Parmer™ RC-200 Series Stuart Recirculating Cooler
Product Code. 16354296
Click to view available options
Cooling Capacity:
450 Watts (at 10°C)
Unit Size:
Each
This item is not returnable. Vea la política de devoluciones
Código de producto. 16354296
Marca: Cole-Parmer™ 1215277
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones
This RC-200 Series Stuart Recirculating Cooler provides powerful cooling in a compact footprint.
  • Optimize accuracy with easy temperature offset calibration
  • Worry-free operation—visual safety alarm indicates overload
  • Easy to read current temperature readout on LED display
  • Reservoir capacity: 3 L
  • 3 year warranty
  • Previous part number SRC5
This reliable, easy-to-use recirculating cooler offers accurate and powerful cooling for an external device. Cooler can achieve temperatures down to -20°C, making it ideal for controlling equipment in temperature-sensitive applications.
Featuring convenient temperature regulation, the LED display features the current temperature, alarm indicator, and press button temperature set point control. An easy-to-use calibration function allows you to adjust the temperature display to match a traceable temperature standard.
Use the 9 mm ID hose barb inlet/outlet ports to connect to a variety of external equipment. A dedicated drain port allows for easy emptying and cleaning.
Note: Do not use water in cooler for low temperatures. Requires use of an antifreeze-type liquid such as ethylene glycol solution.
Please Note: Effective June 2022, the Stuart Recirculating Cooler Model number SRC5 has now been rebranded Cole-Parmer under Model number RC-200-3. All the same great features and specifications, just with a new and improved look.
Especificaciones
Capacity (English) 0.8 gal.
Capacity (Metric) 3 L
Cooling Capacity 450 Watts (at 10°C)
Flow Rate 9 L/min.
Temperature Accuracy ±2°
Temperature (Metric) Maximum 30°C
Dimensions (L x W x H) 44.5 x 20.5 x 54.5 cm
Electrical Requirements 220 VAC
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado