Learn More
Thermo Scientific™ RapidFinder™ Ruminant ID Kit
Click to view available options
Quantity:
48 reactions
Unit Size:
48 reactions
Description
- Enables the simple and reliable detection of ruminant meat in food and feed samples
- The method works by detection of Bos taurus, Ovis aries, Capra hircus, Cervus elaphus, and Capreolus capreolus DNA, which corresponds to DNA from beef, sheep, goat, and deer, by real-time PCR
- Unlike ELISA based methods, when used with the GMO Extraction Kit (4466336) for sample preparation, the RapidFinder™ Ruminant ID Kit detects both raw and processed ruminant meat and provides sensitivity down to 0.01% ruminant DNA
- The included positive control in the kit, which comprises 0.1% ruminant DNA, can be used as a reference to set the threshold of detection of the assay
- Simple-real-time PCR based assay, no electrophoresis needed
- Assay is compatible with Applied Biosystems™ StepOne™, StepOnePlus™, 7500, 7500 Fast, and 7300 real-time PCR systems
Includes
- Ruminant Master Mix (store at -20°C)
- General Master Mix (store at 4°C or -20°C)
- Positive Control (store at -20°C)
Specifications
Specifications
| Content And Storage | Contains 1 tube each of: Ruminant Master Mix (store at -20°C) General Master Mix (store at -20°C upon receipt; 2-8°C after initial use. Store protected from light) Positive Control (store at -20°C) |
| Detection Method | Primer-probe |
| PCR Method | qPCR |
| Reaction Speed | Standard |
| For Use With (Equipment) | Applied Biosystems™ QuantStudio™ 5 Real‑Time PCR Instrument, Applied Biosystems™ 7500 Fast Real‑Time PCR Instrument |
| Kit Type | Ruminant ID Kit |
| Product Line | RapidFinder |
| Quantity | 48 reactions |
| Species | Ruminant |
| For Use With (Application) | Food and Beverage |
| Show More |