Learn More
Invitrogen™ Citrate Synthase Monoclonal Antibody (GT1761)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA517264
Product Code. 15398240
  • 518.00 EUR / 100µL
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, A375, mouse brain, rat brain, zebrafish brain, zebrafish heart. Predicted reactivity: Mouse (94%), Rat (93%), Zebrafish (90%), Xenopus laevis (91%), Pig (95%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a Krebs tricarboxylic acid cycle enzyme that catalyzes the synthesis of citrate from oxaloacetate and acetyl coenzyme A. The enzyme is found in nearly all cells capable of oxidative metabolism. This protein is nuclear encoded and transported into the mitochondrial matrix, where the mature form is found.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Citrate Synthase
Monoclonal
1 mg/mL
PBS with no preservative
O75390, Q7ZVY5, Q8VHF5, Q9CZU6
CS
Recombinant protein encompassing a sequence within the center region of human Citrate synthetase. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse, Rat, Zebrafish
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
GT1761
Unconjugated
CS
2610511A05Rik; 9030605P22Rik; Ahl4; BB234005; Cis; citrate (Si)-synthase; citrate synthase; citrate synthase precursor (EC 4.1.3.7); citrate synthase, mitochondrial; citrate synthetase; Cs; wu:fb58e04; zgc:55507
Mouse
Protein G
RUO
12974, 1431, 170587, 322339
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid