Learn More
Invitrogen™ VAMP1/VAMP2 Monoclonal Antibody (SP10)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517608
This item is not returnable.
View return policy
Description
MA5-17608 targets VAMP/synaptobrevin in ELISA, FACS, and WB applications and shows reactivity with Human samples. The MA5-17608 immunogen is human Synaptobrevin 1 (aa 1-118) and Synaptobrevin 2(aa 33-96).
Vesicle-associated membrane protein 2 is essential for cAMP-regulated exocytosis. VAMP (also know as synaptobrevin) is part of the synaptic vesicle docking and fusion complex and plays a central role in neuroexocytosis. Two VAMP (vesicle-associated membrane protein) isoforms are expressed in the nervous system and are differently distributed among the specialized parts of the tissue. VAMP-1 and -2 are present in all tissues tested, including kidney, adrenal gland, liver, pancreas, thyroid, heart, and smooth muscle. The two isoforms are differentially expressed in various tissues and their level may depend on differentiation. VAMP-1 is restricted to exocrine pancreas and to kidney tubular cells, whereas VAMP-2 is the predominant isoform present in Langerhans islets and in glomerular cells. Both isoforms show a patchy vesicular intracellular distribution in confocal microscopy. Evidence for the importance of neuronal VAMPproteins in the physiology of all cells is indicated.
Specifications
| VAMP1/VAMP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P23763, P63027 | |
| VAMP1, VAMP2 | |
| Human Synaptobrevin 1 (aa 1-118) and Synaptobrevin 2(aa 33-96). | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
| ELISA, Flow Cytometry, Western Blot | |
| SP10 | |
| Unconjugated | |
| VAMP1 | |
| RATVAMPB; RATVAMPIR; SPAX1; SYB; SYB1; SYB2; synaptobrevin 1; synaptobrevin 2; Synaptobrevin 2 (vesicle-associated membrane protein VAMP-2); synaptobrevin-1; Synaptobrevin-2; Vamp1; VAMP-1; VAMP2; VAMP-2; vesicle associated membrane protein 1; vesicle associated membrane protein 2; Vesicle-associated membrane protein (synaptobrevin 2); vesicle-associated membrane protein 1; vesicle-associated membrane protein 1 (synaptobrevin 1); vesicle-associated membrane protein 2; vesicle-associated membrane protein 2 (synaptobrevin 2) | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6843, 6844 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |