Learn More
IKA™ RW 28 Digital Overhead Stirrer
GREENER_CHOICE
Product Code. 14821451 Shop All IKA Products
Change view
Click to view available options
Speed:
60 to 1400 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed unitSize
14821451 60 to 1400 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 14821451 Supplier IKA™ Supplier No. 0005040000
Only null left
Add to basket
Add to basket
Use this Powerful, mechanically controlled laboratory stirrer for highly viscous applications with quantities up to 80L (H2O). RW 28 Digital Overhead Stirrers are suitable for intensive mixing in laboratories and pilot plants.
  • Equipped with a protection for overheating of the motor by means of self-locking temperature limiter 
  • To secure bowls, a stirring shaft protection and clamping fixture are optionally available
  • Digital speed display
  • Infinitely adjustable speed from 60 to 1400 rpm in two speed ranges
  • Push-through agitator shafts
  • Overload protection
  • Error code display
  • Robust, ergonomic design
  • Quiet operation
  • With constant power-drive
TRUSTED_SUSTAINABILITY
Specifications
Speed 60 to 1400 rpm
Capacity 80 L
Display LED
Voltage 240 V
Wattage 220 w
Height (Metric) 346 mm
Width (Metric) 123 mm
Electrical Requirements 240V, 50/60Hz, 220w
Frequency 50/60 Hz
Material Alu-Cast Coating/Thermoplastic Polymer
Model RW 28 Digital
Temperature Range (Metric) 5°C to 40°C
Torque 900 Ncm
Viscosity 50,000 mPas