Learn More
Ziehl Fuchsine-Carbol Solution, Chem-Lab
missing translation for 'orderingAttributeHoverText'
Quantity:
2.5 L
missing translation for 'unitSize'
2.5 L
Beschreibung
Initially, fuchsin-carbol stains every cell. When the staining solution is combined with acid alcohol (usually hydrochloric acid in ethanol or IPA), the stain is removed from non-acid-fast bacteria, since they do not have a waxy lipid layer that resists decolorization. A methylene blue counter stain can then applied, which is picked up by non-acid-fast bacteria. When viewed under the microscope, the acid-fast bacteria appear red, while other bacteria appear blue.
Spezifikation
Spezifikation
| Melting Point | 0°C |
| Boiling Point | 100°C |
| CAS | 632-99-5, 108-95-2, 64-17-5, 7732-18-5 |
| Concentration | Fuchsine, Phenol and Ethanol in Water |
| Solubility Information | Soluble in water |
| Odor | Weak |
| Density | 0.97g/mL |
| Color | Red |
| pH | ±7 |
| Quantity | 2.5 L |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- Fuchsine acc. Ziehl solution
- Danger
- Acute toxicity Category 4
- Flammable liquid Category 3
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 1 B
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H341-Suspected of causing genetic defects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Fuchsine basic, Phenol, Ethanol
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur