Learn More
Brady™ B-422 Polyester Thermal Transfer Permanent Labels
Product Code. 11562715
Click to view available options
Height (Metric):
5.1 mm
9.5 mm
12.7 mm
25.4 mm
31.8 mm
50.8 mm
76.2 mm
Vertical Repeat (Metric):
7.6 mm
12.7 mm
15.2 mm
28.6 mm
34.9 mm
54 mm
79.4 mm
Width (Metric):
12.7 mm
25.4 mm
38.1 mm
50.8 mm
69.9 mm
76.2 mm
101.6 mm
Unit Size:
1 roll
Product Code. 11562715
Brand: Brady™ 014941
Only null left
Add to basket
Add to basket
Printable, permanent polyester labels for thermal transfer printers.
  • High adhesion
  • Acrylic adhesive labels
  • Heat resistant
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance UL and CSA Approved
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Gloss
Format Die-cut
Material Polyester (B-422)
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Asset Tracking, Barcode Labeling, Equipment Labeling, General Identification
For Use With (Equipment) BBP81, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
Model THT-7-422-3
No. per Pack 3000 Label(s) / Roll
Quantity 3000 Labels
Shape Rectangle
Size 69.85 mm (W) x 31.75 mm (H)
Temperature Range (English) -40°F to 212°F
Temperature Range (Metric) -40°C to +100°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Vertical Repeat (Metric) 34.9 mm
Dimensions (LxW) 69.85 mm (W) x 31.75 mm (H)
Length (English) 1.25 in.
Width (English) 2.75 in.
Width (Metric) 69.9 mm
Width (English) Printable Area 69.85 mm
Width (Metric) Printable Area 69.9 mm
Height (English) 1.25 in.
Height (Metric) 31.8 mm
Height (Metric) Printable Area 31.8 mm
Alternate Part Number THT-7-422-3