Learn More
Brady™ B-430 Polyester Thermal Transfer Labels for Laminat™ Carriers and Tags
Product Code. 11529883
Click to view available options
Height (Metric):
5.1 mm
11.1 mm
21 mm
74 mm
124 mm
129 mm
Labels per Row:
1
2
3
5
Vertical Repeat (Metric):
7.9 mm
14.1 mm
24.3 mm
77 mm
127 mm
133.4 mm
Width (Metric):
11 mm
21 mm
37 mm
39 mm
49 mm
Unit Size:
1 roll
Product Code. 11529883
Brand: Brady™ 620362
Only null left
Add to basket
Add to basket
Polyester labels suitable for printing with Thermal Transfer Printers.
  • Specifically designed to fit Laminat™ Carriers and Tags
  • For smooth surfaces
  • Permanent acrylic adhesive
Specifications
Certifications/Compliance UL Recognized
Adhesive Type Acrylic, Permanent
Color Transparent
Finish Gloss
Format Die-cut
Material Polyester (B-430)
Product Type Label
Sterility Non-sterile
For Use With (Application) Asset Tracking, Equipment Labeling, Data, Telecommunications and Electrical Labeling, General Identification, Inventory Labeling
For Use With (Equipment) BBP11, BBP12, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
Horizontal Repeat (Metric) 40 mm
Labels per Row 2
Quantity 2000 Labels
Shape Rectangle
Temperature Range (English) -94°F to +212°F
Temperature Range (Metric) -70°C to +100°C
Thickness (English) 0.003 in.
Thickness (Metric) 0.076 mm
Vertical Repeat (Metric) 24.3 mm
Dimensions (LxW) 37.00 mm (W) x 21.00 mm (H)
Length (English) 0.83 in.
Width (English) 1.46 in.
Width (Metric) 37 mm
Width (Metric) Printable Area 37 mm
Height (English) 0.83 in.
Height (Metric) 21 mm
Alternate Part Number THTTL-4-430