Learn More
Thermo Scientific™ Acclaim™ HILIC-10 HPLC Columns
Product Code. 11312053 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
10 mm
150 mm
Diameter (Metric):
2.1 mm
3 mm
4.6 mm
Particle Size:
3 μm
5 μm
Unit Size:
Each
Pack of 2
Product Code. 11312053
Brand: Thermo Scientific™ 074262
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use this HILIC column to retain and separate highly polar molecules that are unretained by reversed-phase chromatography.
Separate highly hydrophilic molecules by hydrophilic interaction liquid chromatography using Thermo Scientific™ Acclaim™ HILIC-10 LC Columns. These columns retain and separate highly polar molecules that are unretained by reversed-phase chromatography. Their high-purity, spherical, porous silica base is covalently modified with a proprietary hydrophilic layer. This chemistry enables the use of 5–20% aqueous mobile phase while maintaining affinity for polar analytes. Sample throughput is increased without the need to evaporate and reconstitute samples dissolved in nonaqueous solvents.
Rugged and Reproducible Separations of Highly Polar Compounds:
  • Unique selectivity, complementary to reversed-phase columns
  • Enhanced sensitivity for MS detection
  • Hydrolytically stable
  • Rugged column packing
  • Broad application range, including separation of hydrophilic drugs and drug metabolites
Acclaim HILIC-10 LC columns are available in 3μm particle size and a variety of column formats: 4.6×150mm, for high-throughput routine analysis; 3.0×150mm, for high-throughput analysis with reduced solvent consumption; and 2.1×150mm, for LC-MS analysis.
TRUSTED_SUSTAINABILITY
Specifications
Column Format Guard Cartridge, 2 Pk.
Diameter (Metric) 4.6 mm
For Use With LC/MS
Length (Metric) 10 mm
Max. Pressure 6000 psi (415 bar)
Particle Size 5 μm
pH 2 to 8
Pore Size 120 Å
Surface Area 300 m²/g
Temperature 70°C
USP Type N/A
Stationary Phase Proprietary
Product Line Acclaim
Packing Material Silica
Carbon Load 8%
Column Type HILIC
Endcapped Yes
Phase HILIC
Type Guard Cartridge, 2 Pk.
Show More Show Less
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.