Learn More
Thermo Scientific Chemicals New methylene Blue N, pure
Product Code. 10742592 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Product Code. 10742592
Brand: Thermo Scientific Chemicals undefined
Request Bulk or Custom Format
CAS: 1934-16-3 | C18H22ClN3S | 347.905 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 1934-16-3
Dye Content ca 70%
Molecular Formula C18H22ClN3S
MDL Number MFCD00135878
Beilstein 27, IV, 5163
Synonym Basic Blue 24
InChI Key SQHOAFZGYFNDQX-UHFFFAOYSA-N
Infrared Spectrum Authentic
SMILES CCNC1=CC2=C(C=C1C)N=C3C=C(C(=[NH+]CC)C=C3S2)C.[Cl-]
IUPAC Name ethyl-[7-(ethylamino)-2,8-dimethylphenothiazin-3-ylidene]azanium;chloride
Molecular Weight (g/mol) 347.905
PubChem CID 73518
Formula Weight 347.9
Grade Pure
Packaging Glass bottle
Color Brown to Green
Quantity 2.5 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material New methylene Blue N, 85%
Show More Show Less
RUO – Research Use Only