Learn More
Thermo Scientific™ CellSensor™ NFAT bla HEK 293T Cell Line
Description
The CellSensor™ NFAT-bla HEK 293T cell line contains a beta-lactamase reporter gene under the control of the Nuclear Factor of Activated T Cells (NFAT) response element stably integrated into HEK 293T cells. Fluorescence activated cell sorting (FACS) was used to isolate clones responsive to stimulation of the NFAT pathway by PMA (Phorbol 12-Myristate 13-Acetate) and Thapsagargin (Thaps). The cell line was validated for DMSO tolerance, cell number, stimulation time, substrate loading conditions, and EC50 concentration of Thaps at fixed concentration of PMA. The CellSensor™ NFAT-bla HEK 293T cell line responds to agonist treatment as expected from the literature and can be adapted for high-throughput screening (including coupling of receptors to the NFAT pathway) for agonists or antagonists with compound libraries. Candidate drugs can also be tested for dose response against this cell line. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Description | CellSensor™ NFAT bla HEK 293T Cell Line, 1Vial, ∽ 2,000,000 cells/vial at 2,000,000 cells/mL Quantity of Cells, NFAT bla HEK 293T Cell Line, Fluorescent Detection Method, CellSensor™ Druggable Target, Adherent Growth Properties, Liquid Nitrogen Storage |
| Detection Method | Fluorescent |
| For Use With (Application) | Pharma and biopharma, drug discovery and development, target and lead identification and validation, kinase biology, kinase pathway analysis, pathway biology, cellular pathway analysis assays, cellSensor™ cellular pathway assays |
| Includes | beta-lactamase reporter gene under the control of the nuclear factor of activated T cells (NFAT) response element stably integrated into HEK 293T cells |
| Product Line | CellSensor |
| Quantity | 1 Vial |
| Sample Type | NFAT-bla HEK 293T |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.