Learn More
BRAND™ Centrifuge Tubes
Product Code. 10517651 Shop All Brand Products
Click to view available options
Capacity (Metric):
75 mL
Unit Size:
Pack of 20
Product Code. 10517651
Brand: BRAND™ 115352
Only null left
Add to basket
Add to basket
Beaded rim, without stopper
  • Beaded rim, without stopper
  • Withstands RCF up to 4500
  • Autoclavable (121 °C)
  • 35 x 100 mm
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 75 mL
Material Polypropylene
No. per Pack 20
Shape Cylindrical
Closure Included No
Product Type Centrifuge Tube