Learn More
BRAND™ Dispenser, tilt measure without bottle
Product Code. 10012392 Shop All Brand Products
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
Unit Size:
Each
Product Code. 10012392
Brand: BRAND™ 43088
Tilt dispenser is best suited for dispensing suspensions.
  • Dispenser head
  • Calibrated to deliver (TD, Ex)
  • Inscriptions in green enamel
  • Particularly well-suited for dispensing suspensions
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass 3.3
Description Without bottle
Capacity (Metric) 100 mL