missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ VIC™ Dye Calibration Plate, 384-well

Product Code. 15820393 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 x 384 well plate
Unit Size:
Each
Product Code. Quantity unitSize
15820393 1 x 384 well plate Each
1 options
This item is not returnable. View return policy

Product Code. 15820393

Brand: Applied Biosystems™ 4432452

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The 384-Well Dye Calibration Plate (VIC™) is used to maintain your real-time PCR instrument with 384-well block

  • The 384-Well Dye Calibration Plate (VIC™) is used to maintain your real-time PCR instrument with 384-well block.
  • Dye calibration should be run after lamp change.
  • Please see your instrument user guide for full instructions on use.
TRUSTED_SUSTAINABILITY

Specifications

Format 384-well Dye Calibration Plate
Content And Storage Store at –20°C ± 5°C.

Calibrated Dye VIC
For Use With (Equipment) ViiA™ 7 Dx System, QuantStudio™ Dx
Product Type Dye Calibration Plate
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Dry Ice
Quantity 1 x 384 well plate
Calibration Type Spectral Calibration
PCR Method qPCR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.