Learn More
Motic™ VI-LED Vertical Illuminator, 3W
Click to view available options
For Use With (Equipment):
SMZ-161 and SMZ-171 Microscopes
Unit Size:
Each
Description
- On-axis observation and documentation.
- Specially designed for Motic SMZ-161 and SMZ-171.
- True on-axis observation of high-resolution, high-contrast, shadow-free images.
- Eliminates angular specimen view characteristic of traditional stereoscopes.
- Ideal for observing particularly smooth, specular surfaces and highly reflective specimens such as integrated circuits, semiconductor wafers, polished metal parts, solder balls, or magnetic recording heads.
- Directs light from the 3W LED down through the objective onto the specimen.
- Universal power supply (100-240V).
- Environmentally-friendly with long-lasting 3W LED bulb and lamp life up to 20,000 hours.
Specifications
Specifications
| Type | Vertical, LED, 3 w |
| Description | Universal power supply (100-240V) |
| For Use With (Equipment) | SMZ-161 and SMZ-171 Microscopes |
| Voltage | 100/240 V |