Learn More
Motic™ VI-LED Vertical Illuminator, 3W
Product Code. 15475452
Click to view available options
For Use With (Equipment):
SMZ-161 and SMZ-171 Microscopes
Unit Size:
Each
Product Code. 15475452
Brand: Motic™ 1101010100151
Only null left
Add to basket
Add to basket
Specially designed for SMZ-161 and SMZ-171 Microscopes. Motic™ VI-LED Vertical Illuminator, 3W uses a groundbreaking optical and illumination system to enable on-axis observation and documentation.
  • On-axis observation and documentation.
  • Specially designed for Motic SMZ-161 and SMZ-171.
  • True on-axis observation of high-resolution, high-contrast, shadow-free images.
  • Eliminates angular specimen view characteristic of traditional stereoscopes.
  • Ideal for observing particularly smooth, specular surfaces and highly reflective specimens such as integrated circuits, semiconductor wafers, polished metal parts, solder balls, or magnetic recording heads.
  • Directs light from the 3W LED down through the objective onto the specimen.
  • Universal power supply (100-240V).
  • Environmentally-friendly with long-lasting 3W LED bulb and lamp life up to 20,000 hours.
Specifications
Type Vertical, LED, 3 w
Description Universal power supply (100-240V)
For Use With (Equipment) SMZ-161 and SMZ-171 Microscopes
Voltage 100/240 V