missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Verteporfin, MedChemExpress
Description
Verteporfin (CL 318952) is a photosensitizer for photodynamic therapy to eliminate the abnormal blood vessels in the eye associated with conditions such as age-related macular degeneration. Verteporfin is a YAP inhibitor which disrupts YAP-TEAD interactions. Verteporfin induces cell apoptosis. Verteporfinis an autophagy inhibitor that blocks autophagy at an early stage by inhibiting autophagosome formation.
Specifications
Specifications
| Chemical Name or Material | Verteporfin |
| CAS | 129497-78-5 |
| Color | Brown |
| Purity Grade Notes | Research |
| Molecular Formula | C41H42N4O8 |
| Quantity | 5 mg |
| Synonym | CL 318952 |
| Solubility Information | DMSO : 5 mg/mL (6.96 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | C[C@]1(/C2=C/C(N3)=C4C)C(C(/C=C(C(C)=C/5C=C)\NC5=C/C6=N/C(C(CCC(OC)=O)=C6C)=C\C3=C4CCC(O)=O)=N2)=CC=C(C(OC)=O)[C@H]1C(OC)=O.C[C@]7(/C8=C/C(N9)=C%10C)C(C(/C=C(C(C)=C/%11C=C)\NC%11=C/C%12=N/C(C(CCC(O)=O)=C%12C)=C\C9=C%10CCC(OC)=O)=N8)=CC=C(C(OC)=O)[C@H]7C(OC)=O |
| Molecular Weight (g/mol) | 718.79 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?